Learn More
Medchemexpress LLC 2-Amino-5-fluorophenol | 53981-24-1 | 127.12 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW00734550G
Encompass_Preferred
2-Amino-5-fluorophenol is an isomer of aminofluorophenol and a HONO generator. It reacts with nitrogen dioxide (NO2) through two pathways to form nitrous acid (HONO): hydrogen abstraction from the hydroxyl group and hydrogen abstraction from the amino group. Both the amino and hydroxyl groups are involved in the reaction in the transition state.
  • An isomer of aminofluorophenol
  • Functions as a HONO generator
  • Reacts with nitrogen dioxide (NO2) to form nitrous acid (HONO)
  • Involves both amino and hydroxyl groups in the reaction
Catalog No. 50-004-60239