Learn More
2-Amino-5-fluorophenol, 97%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
2-Amino-5-fluorophenol is a fluoroaniline metabolite. 2-Amino-5-fluorophenol is used as organic intermediate. To synthesize the intermediate (2-amino-5-fluorophenol-N, N, O-triacetic acid trimethyl ester) of indicator which used in the determination Mg2+ by NMR, three feasible reactions were experimentally carried out, such as alkaline hydrolysis, catalytic hydrogenation and esterification.
Solubility
Slightly soluble in water.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Refrigerated.
Specifications
Specifications
| CAS | 53981-24-1 |
| Molecular Formula | C6H6FNO |
| Quantity | 250mg |
| MDL Number | MFCD00671759 |
| Sensitivity | Air sensitive |
| Synonym | 4-fluoro-2-hydroxyaniline, phenol, 2-amino-5-fluoro, 2-amino-5-fluoro-phenol, 5-fluoro-2-aminophenol, 2-hydroxy-4-fluoroaniline, pubchem2828, acmc-1avob, 2-amino-5-flourophenol, 2-amino-5-fluor-phenol, 2-amino-5-fluoro phenol |
| Solubility Information | Slightly soluble in water. |
| InChI Key | IIDUNAVOCYMUFB-UHFFFAOYSA-N |
| SMILES | C1=CC(=C(C=C1F)O)N |
| IUPAC Name | 2-amino-5-fluorophenol |
| Show More |
Safety and Handling
| Hazard Category | H302+H312-H315-H319-H335 |
| Hazard Statement | GHS H Statement H302-H312-H315-H319-H335 Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Keep cold; Store under Argon |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.