Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-fluorophenol 97.0+%, TCI America™
SDP

Catalog No. A23005G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23005G 5 g
A23001G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23005G Supplier TCI America Supplier No. A23005G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 53981-24-1
Molecular Formula C6H6FNO
Molecular Weight (g/mol) 127.118
MDL Number MFCD00671759
InChI Key IIDUNAVOCYMUFB-UHFFFAOYSA-N
Synonym 4-fluoro-2-hydroxyaniline, phenol, 2-amino-5-fluoro, 2-amino-5-fluoro-phenol, 5-fluoro-2-aminophenol, 2-hydroxy-4-fluoroaniline, pubchem2828, acmc-1avob, 2-amino-5-flourophenol, 2-amino-5-fluor-phenol, 2-amino-5-fluoro phenol
PubChem CID 185763
IUPAC Name 2-amino-5-fluorophenol
SMILES C1=CC(=C(C=C1F)O)N

Specifications

Quantity 5 g
Formula Weight 127.12
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-fluorophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.