Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Amino-5-methylbenzoic acid | 2941-78-8 | 151.16 | 500 G
SDP

Catalog No. 5000447377
Encompass_Preferred

2-Amino-5-methylbenzoic Acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is an impurity of cetilistat.

  • Biochemical reagent
  • Biological material or organic compound
  • Used for life science related research
  • Off-white to light yellow solid appearance
  • Purity of 99.8%
  • Molecular weight of 151.16
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-47377 Supplier Medchemexpress LLC Supplier No. HYW002036500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Amino-5-methylbenzoic Acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is an impurity of cetilistat.

  • Biochemical reagent
  • Biological material or organic compound
  • Used for life science related research
  • Off-white to light yellow solid appearance
  • Purity of 99.8%
  • Molecular weight of 151.16
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.