Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-methylphenol, 98%

Catalog No. p-7027084
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2356009 10 g
AAB2356018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2356009 Supplier Thermo Scientific Chemicals Supplier No. B2356009
Only null left
Add to Cart

CAS: 2835-98-5 | C7H9NO | 123.155 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2835-98-5
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD00007693
InChI Key HCPJEHJGFKWRFM-UHFFFAOYSA-N
Synonym 6-amino-m-cresol, 2-hydroxy-4-methylaniline, phenol, 2-amino-5-methyl, 4-amino-3-hydroxytoluene, 2-hydroxy-p-toluidine, unii-qcg4es2a26, 6-amino-3-cresol, 3-methyl-6-aminophenol, 6-amino-3-methylphenol, qcg4es2a26
PubChem CID 76082
IUPAC Name 2-amino-5-methylphenol
SMILES CC1=CC(=C(C=C1)N)O

Specifications

Melting Point 155°C to 159°C
Quantity 10 g
Beilstein 386144
Formula Weight 123.16
Percent Purity 98%
Chemical Name or Material 2-Amino-5-methylphenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 220-620-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.