Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Amino-5-methylpyrazine | 5521-58-4 | 99.7% | C5H7N3 | 1 G
SDP

Catalog No. 5000433781
Encompass_Preferred

2-Amino-5-methylpyrazine is an intermediate chemical compound primarily used in the synthesis of LPAR1 antagonists. This product is intended for research purposes only and not for patient use.

  • Utilized as an intermediate for LPAR1 antagonist synthesis.
  • Purity of 99.7%.
  • Appears as a light brown to yellow solid.
  • Highly soluble in DMSO.
  • Store at 4°C, protected from light.

Catalog No. 50-004-33781 Supplier Medchemexpress LLC Supplier No. HY652551G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Amino-5-methylpyrazine is an intermediate chemical compound primarily used in the synthesis of LPAR1 antagonists. This product is intended for research purposes only and not for patient use.

  • Utilized as an intermediate for LPAR1 antagonist synthesis.
  • Purity of 99.7%.
  • Appears as a light brown to yellow solid.
  • Highly soluble in DMSO.
  • Store at 4°C, protected from light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.