Learn More
Medchemexpress LLC 2-Amino-5-methylpyrazine | 5521-58-4 | 99.7% | C5H7N3 | 5 G
SDP
Catalog No. 5000434765
Encompass_Preferred
2-Amino-5-methylpyrazine is a chemical intermediate designed for research applications. It is utilized in the synthesis of LPAR1 antagonists. This product presents as a solid, ranging in color from light brown to yellow, and boasts a high purity of 99.72%.
  • Functions as an intermediate for chemical synthesis.
  • Enables the synthesis of LPAR1 antagonists.
  • Exhibits a high purity.
  • Available as a solid form.
  • Displays a light brown to yellow coloration.
  • Highly soluble in dimethyl sulfoxide.
  • Features multiple dissolution protocols for in vivo studies.
  • Intended exclusively for research purposes.
Catalog No. 50-004-34765 Supplier Medchemexpress LLC Supplier No. HY652555G
2-Amino-5-methylpyrazine is a chemical intermediate designed for research applications. It is utilized in the synthesis of LPAR1 antagonists. This product presents as a solid, ranging in color from light brown to yellow, and boasts a high purity of 99.72%.
  • Functions as an intermediate for chemical synthesis.
  • Enables the synthesis of LPAR1 antagonists.
  • Exhibits a high purity.
  • Available as a solid form.
  • Displays a light brown to yellow coloration.
  • Highly soluble in dimethyl sulfoxide.
  • Features multiple dissolution protocols for in vivo studies.
  • Intended exclusively for research purposes.