Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-methylthio-1,3,4-thiadiazole, 99%, Thermo Scientific™

Catalog No. AC329700250
Change view
Click to view available options
:
25g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC329700250 25g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC329700250 Supplier Thermo Scientific Chemicals Supplier No. 329700250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5319-77-7
CAS Min % 98.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C3H5N3S2
Quantity 25g
MDL Number MFCD00175752
Synonym 5-methylthio-1,3,4-thiadiazol-2-amine, 2-amino-5-methylthio-1,3,4-thiadiazole, 1,3,4-thiadiazol-2-amine, 5-methylthio, 1,3,4-thiadiazole, 2-amino-5-methylthio, 5-methylsulfanyl-1,3,4-thiadiazol-2-amine, 2-amino-5-methylmercapto-1,3,4-thiadiazole, 5-methylthio-1,3,4-thiadiazole-2-ylamine, 4-27-00-08165 beilstein handbook reference, 2-amino-5-methylthio-1,3,4-thidiazole
InChI Key PCLAZAJARAIGGD-UHFFFAOYSA-N
SMILES CSC1=NN=C(N)S1
IUPAC Name 5-(methylsulfanyl)-1,3,4-thiadiazol-2-amine
Molecular Weight (g/mol) 147.21
PubChem CID 21390
Formula Weight 147.23
Percent Purity ≥98.5%
Chemical Name or Material 2-Amino-5-methylthio-1, 3, 4-thiadiazole, 99%
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECSNumber XI3540000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.