Learn More
Medchemexpress LLC 2-amino-5-methylthiophene-3-carbonitrile | 138564-58-6 | MFCD01626055 | 99.8% | 138.19 g·mol⁻¹ | C6H6N2S | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0157735G
Encompass_Preferred
2-Amino-5-methylthiophene-3-carbonitrile is a thiophene-derived building block supplied for research and synthetic chemistry. It is commonly used as an intermediate in heterocycle construction and has documented use in the synthesis of olanzapine. The material is provided as a solid or as a ready-to-use solution in DMSO and is supported by analytical and safety documentation.
  • High purity suitable for research applications (≈99.8%).
  • Available as solid and as a 10 mM solution in DMSO.
  • Supplied with analytical documentation and safety data.
  • Molecular formula C6H6N2S and molecular weight 138.19 g·mol⁻¹.
  • Useful as a building block for heterocycle synthesis and pharmaceutical intermediates.
  • Multiple pack sizes available from small-scale to bulk options.
Catalog No. 50-003-00520