Learn More
Medchemexpress LLC 2-Amino-5-nitrobenzophenone | 1775-95-7 | 98.3% | 50 G
SDP
Catalog No. 5000410339
Encompass_Preferred
2-Amino-5-nitrobenzophenone (Ro-7-19990) is a substance resulting from the degradation of benzodiazepines. It is suitable for use in both research and forensic science applications.
  • Degradation product of benzodiazepines.
  • Applicable in research settings.
  • Suitable for forensic applications.
Catalog No. 50-004-10339 Supplier Medchemexpress LLC Supplier No. HYB212050G
2-Amino-5-nitrobenzophenone (Ro-7-19990) is a substance resulting from the degradation of benzodiazepines. It is suitable for use in both research and forensic science applications.
  • Degradation product of benzodiazepines.
  • Applicable in research settings.
  • Suitable for forensic applications.