Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-nitropyrimidine 98.0+%, TCI America™
SDP

Catalog No. A20311G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A20311G Supplier TCI America Supplier No. A20311G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3073-77-6
Molecular Formula C4H4N4O2
Molecular Weight (g/mol) 140.102
MDL Number MFCD00006103
InChI Key SSHFCFRJYJIJDV-UHFFFAOYSA-N
Synonym 2-amino-5-nitropyrimidine, 2-pyrimidinamine, 5-nitro, 5-nitro-pyrimidin-2-ylamine, 5-nitro-2-pyrimidinamine, 5-nitropyrimidine-2-ylamine, 5-nitropyrimidin-2-amin, pubchem6898, amino-5-nitropyrimidine, 2-amino-5-nitropyrimdin, acmc-1cjp5
PubChem CID 76490
IUPAC Name 5-nitropyrimidin-2-amine
SMILES C1=C(C=NC(=N1)N)[N+](=O)[O-]

Specifications

Melting Point 236°C
Quantity 1 g
Formula Weight 140.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-nitropyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.