Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-phenyl-3-furonitrile, 95%, Thermo Scientific™

Catalog No. p-3880515
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AW01254DA 1 g Amber Glass Bottle
AW01254EA 10 g Amber Glass Bottle
AW01254EB 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AW01254DA Supplier Thermo Scientific Chemicals Supplier No. AW01254DA
Only null left
Add to Cart

Chemical Identifiers

CAS 14742-32-6
Molecular Formula C11H8N2O
Molecular Weight (g/mol) 184.20
MDL Number MFCD00463472
InChI Key UWTDTJWGYWUILP-UHFFFAOYSA-N
Synonym 2-amino-5-phenyl-3-furonitrile, 2-amino-3-cyano-5-phenylfuran, 2-amino-5-phenyl-3-furancarbonitrile, 3-furancarbonitrile, 2-amino-5-phenyl, maybridge1_000452, 2-amino-5-phenyl-furan-3-carbonitrile, 3-furancarbonitrile,2-amino-5-phenyl, 2-azanyl-5-phenyl-furan-3-carbonitrile
PubChem CID 693961
IUPAC Name 2-amino-5-phenylfuran-3-carbonitrile
SMILES NC1=C(C=C(O1)C1=CC=CC=C1)C#N

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 184.2
Percent Purity 95%
Chemical Name or Material 2-Amino-5-phenyl-3-furonitrile
Hazard Category GHS Signal Word: Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.