Learn More
Medchemexpress LLC HY-W002820 500mg Medchemexpress, 2-Amino-5-phenylpyridine CAS:33421-40-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW002820500MG
Encompass_Preferred
Medchemexpress, HY-W002820 500mg 2-Amino-5-phenylpyridine CAS:33421-40-8 2-Amino-5-phenylpyridine is a mutagenic heterocyclic aromatic amine that is formed by pyrolysis of phenylalanine in proteins. 2-Amino-5-phenylpyridine is in broiled sardines and is considered as potentially carcinogenic. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3213