Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+/-)-2-Amino-5-phosphonopentanoic acid, 99%, Thermo Scientific™

Catalog No. AAJ64210MF
Change view
Click to view available options
Quantity:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64210MF 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64210MF Supplier Thermo Scientific Chemicals Supplier No. J64210MF

missing translation for 'validMainframeMsg'

A potent NMDA antagonist

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 76326-31-3
Molecular Formula C5H12NO5P
Quantity 50mg
MDL Number MFCD00010515
Beilstein 2446389
Sensitivity Light Sensitive
Synonym 5-phosphononorvaline, 2-amino-5-phosphovaleric acid, dl-ap5, dl-2-amino-5-phosphonovaleric acid, dl-2-amino-5-phosphonopentanoic acid, 2-amino-5-phosphonovaleric acid, 2-amino-5-phosphopentanoic acid, 2-amino-5-phosphonovalerate, 2-amino-5-phosphonopentanoate, dl-apv
InChI Key VOROEQBFPPIACJ-UHFFFAOYSA-N
SMILES C(CC(C(=O)O)N)CP(=O)(O)O
IUPAC Name 2-amino-5-phosphonopentanoic acid
Molecular Weight (g/mol) 197.127
PubChem CID 1216
ChEBI CHEBI:138644
Formula Weight 197.13
Percent Purity 99%
Chemical Name or Material (+/-)-2-Amino-5-phosphonopentanoic acid
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.