Learn More
Medchemexpress LLC 2-amino-6-bromopyridine | 19798-81-3 | MFCD00137843 | 99.8% | 173.01 | C5H5BrN2 | 50g
SDP
Catalog No. 5000267525
Encompass_Preferred
2-Amino-6-bromopyridine is an intermediate 2-Amino-6-bromopyridine is also a PqsR ligand with a Kd value of 6 8 M in SPR assay 2-Amino-6-bromopyridine shows weak antagonistic activity 2-Amino-6-bromopyridine can be used in the synthesis of JAK2 inhibitors MSK1 inhibitors and in the research of Pseudomonas aeruginosa infection[1][2][3][4]
Catalog No. 50-002-67525 Supplier Medchemexpress LLC Supplier No. HYY104950G
2-Amino-6-bromopyridine is an intermediate 2-Amino-6-bromopyridine is also a PqsR ligand with a Kd value of 6 8 M in SPR assay 2-Amino-6-bromopyridine shows weak antagonistic activity 2-Amino-6-bromopyridine can be used in the synthesis of JAK2 inhibitors MSK1 inhibitors and in the research of Pseudomonas aeruginosa infection[1][2][3][4]