Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-6-chloro-3-formylchromone 98.0+%, TCI America™
SDP

Catalog No. A27431G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27431G 1 g
A27435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27431G Supplier TCI America Supplier No. A27431G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 68301-77-9
Molecular Formula C10H6ClNO3
Molecular Weight (g/mol) 223.61
MDL Number MFCD00139140
InChI Key GCFVPLUXLQEQJX-UHFFFAOYSA-N
Synonym 2-Amino-6-chlorochromone-3-carboxaldehyde, 2-Amino-6-chloro-4-oxo-4H-1-benzopyran-3-carboxaldehyde
PubChem CID 689030
IUPAC Name 2-amino-6-chloro-4-oxo-4H-chromene-3-carbaldehyde
SMILES NC1=C(C=O)C(=O)C2=CC(Cl)=CC=C2O1

Specifications

Melting Point 290°C
Quantity 1 g
Formula Weight 223.61
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-6-chloro-3-formylchromone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.