Learn More
Medchemexpress LLC 2-amino-6-chlorobenzoic acid | 2148-56-3 | MFCD00051530 | 99.3% | 171.58 | C7H6ClNO2 | 500g
SDP
Catalog No. 5000267377
Encompass_Preferred
2-Amino-6-chlorobenzoic acid is an autocoupling diazotization reagent used for spectrophotometric detection of nitrite ions in water 2-Amino-6-chlorobenzoic acid exhibits good tolerance to various anions under acidic conditions 2-Amino-6-chlorobenzoic acid is commonly used as a reagent in organic synthesis and analytical chemistry[1]
Catalog No. 50-002-67377 Supplier Medchemexpress LLC Supplier No. HYY1132500G
2-Amino-6-chlorobenzoic acid is an autocoupling diazotization reagent used for spectrophotometric detection of nitrite ions in water 2-Amino-6-chlorobenzoic acid exhibits good tolerance to various anions under acidic conditions 2-Amino-6-chlorobenzoic acid is commonly used as a reagent in organic synthesis and analytical chemistry[1]