Learn More
Medchemexpress LLC 2-Amino-6-fluorobenzoic acid | 434-76-4 | 155.13 | 1 ML
SDP
Catalog No. 5000446320
Encompass_Preferred
2-Amino-6-fluorobenzoic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • High purity: 99.99%
  • Recommended storage conditions for powder: -20°C for 3 years, or 4°C for 2 years
  • Suitable for life science research
  • Can be used as a biological material or organic compound
Catalog No. 50-004-46320 Supplier Medchemexpress LLC Supplier No. HYW00113110MM/1ML
2-Amino-6-fluorobenzoic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • High purity: 99.99%
  • Recommended storage conditions for powder: -20°C for 3 years, or 4°C for 2 years
  • Suitable for life science research
  • Can be used as a biological material or organic compound