Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (-)-2-Amino-6-Mercaptopurine Riboside Hydrate

Numéro de catalogue. 8584121G missing translation for 'brandsLinkText'
Encompass
Modifier l'affichage
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
85-841-21G 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 85-841-21G Fournisseur Sigma Aldrich Code fournisseur 8584121G
May include imposed supplier surcharges.
Ajouter au panier
Ajouter au panier

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(-)-2-Amino-6-Mercaptopurine Riboside Hydrate

Spécifications

CAS 559-74-0
Molecular Formula C30H46N2O3
Linear Formula C30H46N2O3
Quantity 5 mg
Synonym (4R)-Hex-5-yn-1-yl 4-((3S,5S,10R,13R,17R)-3-hydroxy-10,13-dimethyl-1,2,3,4,5,7,8,9,10,11,12,13,14,15,16,17-hexadecahydrospiro[cyclopenta[alpha]phenanthrene-6,3'-diazirin]-17-yl)pentanoate
Molecular Weight (g/mol) 482.7 g/mol
missing translation for 'productTitle'
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.