Learn More
Medchemexpress LLC 2-amino-6-methyl-4-propyl-triazolo[1,5-a]pyrimidin-5-one | 27277-00-5 | MFCD06407961 | 99.1% | 207.23 g·mol⁻¹ | C9H13N5O | 250 MG
SDP
Catalog No. 5000247461
Encompass_Preferred
ICI-63197 (CAS 27277-00-5) is a small-molecule phosphodiesterase inhibitor active against PDE3 and PDE4. It exhibits Ki values of 9 μM for PDE3 and 10 μM for PDE4, with substantially lower activity against PDE1 and PDE2. Supplied as a white to off-white solid, the compound is intended for research use in biochemical and pharmacological studies.
  • Potent inhibitor of PDE3 and PDE4 with reported Ki values near 9-10 μM.
  • Selective over PDE1 and PDE2, indicating target specificity.
  • High reported purity suitable for research applications.
  • Available in small-scale quantities for laboratory experiments.
  • Stable as a powder when stored under recommended conditions.
Catalog No. 50-002-47461 Supplier Medchemexpress LLC Supplier No. HY103024250MG
ICI-63197 (CAS 27277-00-5) is a small-molecule phosphodiesterase inhibitor active against PDE3 and PDE4. It exhibits Ki values of 9 μM for PDE3 and 10 μM for PDE4, with substantially lower activity against PDE1 and PDE2. Supplied as a white to off-white solid, the compound is intended for research use in biochemical and pharmacological studies.
  • Potent inhibitor of PDE3 and PDE4 with reported Ki values near 9-10 μM.
  • Selective over PDE1 and PDE2, indicating target specificity.
  • High reported purity suitable for research applications.
  • Available in small-scale quantities for laboratory experiments.
  • Stable as a powder when stored under recommended conditions.