Learn More
Medchemexpress LLC 2-amino-7-(2-furanyl)-7,8-dihydro-5(6H)-quinazolinone | 299442-43-6 | MFCD02026323 | 99.8% | 229.23 | C12H11N3O2 | 100 MG
SDP
Catalog No. 5000246229
Encompass_Preferred
NKY80 is a non-nucleoside quinazolinone small-molecule inhibitor of adenylyl cyclase used in biochemical and pharmacological research to modulate AC catalytic activity, particularly in cardiac and pulmonary studies. For research use only.
  • Potent adenylyl cyclase inhibition with greater affinity for AC5.
  • Well characterized: CAS 299442-43-6; molecular formula C12H11N3O2; molecular weight 229.23 g/mol.
  • High purity (≈99.8%) suitable for biochemical assays.
  • Available in multiple pack sizes, including 100 MG.
  • Soluble in DMSO; follow supplier handling and storage recommendations.
  • Provided as an off-white to light yellow solid.
Catalog No. 50-002-46229 Supplier Medchemexpress LLC Supplier No. HY103195100MG
NKY80 is a non-nucleoside quinazolinone small-molecule inhibitor of adenylyl cyclase used in biochemical and pharmacological research to modulate AC catalytic activity, particularly in cardiac and pulmonary studies. For research use only.
  • Potent adenylyl cyclase inhibition with greater affinity for AC5.
  • Well characterized: CAS 299442-43-6; molecular formula C12H11N3O2; molecular weight 229.23 g/mol.
  • High purity (≈99.8%) suitable for biochemical assays.
  • Available in multiple pack sizes, including 100 MG.
  • Soluble in DMSO; follow supplier handling and storage recommendations.
  • Provided as an off-white to light yellow solid.