Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-amino-7-(furan-2-yl)-7,8-dihydro-6H-quinazolin-5-one | 299442-43-6 | MFCD02026323 | 99.8% | 229.23 g·mol⁻1 | C12H11N3O2 | 50 MG
SDP

Catalog No. 5000245965
Encompass_Preferred

NKY80 is a non-nucleoside quinazolinone inhibitor selective for adenylyl cyclase (AC) type V. It modulates AC catalytic activity in heart and lung tissues and exhibits IC50 values of 8.3 μM (AC V), 132 μM (AC III), and 1.7 mM (AC II). Supplied as a purified research compound for in vitro biochemical and pharmacological studies.

  • Selective adenylyl cyclase type V inhibitor (IC50 8.3 μM).
  • Non-nucleoside quinazolinone chemical class.
  • Approximately 99.8% purity.
  • Molecular weight 229.23 g·mol⁻1; formula C12H11N3O2.
  • Soluble in DMSO; intended for research use only.

Catalog No. 50-002-45965 Supplier Medchemexpress LLC Supplier No. HY10319550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NKY80 is a non-nucleoside quinazolinone inhibitor selective for adenylyl cyclase (AC) type V. It modulates AC catalytic activity in heart and lung tissues and exhibits IC50 values of 8.3 μM (AC V), 132 μM (AC III), and 1.7 mM (AC II). Supplied as a purified research compound for in vitro biochemical and pharmacological studies.

  • Selective adenylyl cyclase type V inhibitor (IC50 8.3 μM).
  • Non-nucleoside quinazolinone chemical class.
  • Approximately 99.8% purity.
  • Molecular weight 229.23 g·mol⁻1; formula C12H11N3O2.
  • Soluble in DMSO; intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.