Learn More
Medchemexpress LLC 2-Amino-8-oxononanoic acid hydrochloride | 2984160-77-0 | 98.0% | C9H18ClNO3 | 100 MG
SDP
Catalog No. 5000355793
Encompass_Preferred
2-Amino-8-oxononanoic acid hydrochloride is the hydrochloride form of 2-Amino-8-oxononanoic acid. It is an amino acid that can be incorporated into proteins in E.coli for genetic studies. This compound is effective for site-specific labeling of proteins with various probes under mild conditions, close to physiological pH.
  • Hydrochloride salt of 2-amino-8-oxononanoic acid
  • Amino acid for genetic incorporation into proteins in E.coli
  • Enables efficient site-specific labeling of proteins
  • Functions effectively under mild conditions near physiological pH
Catalog No. 50-003-55793 Supplier Medchemexpress LLC Supplier No. HY145576B100MG
2-Amino-8-oxononanoic acid hydrochloride is the hydrochloride form of 2-Amino-8-oxononanoic acid. It is an amino acid that can be incorporated into proteins in E.coli for genetic studies. This compound is effective for site-specific labeling of proteins with various probes under mild conditions, close to physiological pH.
  • Hydrochloride salt of 2-amino-8-oxononanoic acid
  • Amino acid for genetic incorporation into proteins in E.coli
  • Enables efficient site-specific labeling of proteins
  • Functions effectively under mild conditions near physiological pH