Learn More
Medchemexpress LLC 2-amino-8-oxononanoic acid hydrochloride | 2984160-77-0 | MFCD11113155 | 98.0% | C9H18ClNO3 | 10MG
SDP
Catalog No. 5000260110
Encompass_Preferred
2-Amino-8-oxononanoic acid hydrochloride is the hydrochloride salt of an amino acid used for site-specific incorporation into proteins expressed in E. coli to enable labeling with probes under mild conditions. It is supplied as a solid with high purity and is available in small milligram pack sizes or as a DMSO solution for biochemical applications.
  • High purity (98.0%) for reproducible results.
  • Hydrochloride salt, solid form for stable storage.
  • Soluble in DMSO (≈50 mg/mL); may require sonication.
  • Stable when stored sealed and kept away from moisture.
  • Suited for site-specific protein labeling in bacterial expression systems.
Catalog No. 50-002-60110 Supplier Medchemexpress LLC Supplier No. HY145576B10MG
2-Amino-8-oxononanoic acid hydrochloride is the hydrochloride salt of an amino acid used for site-specific incorporation into proteins expressed in E. coli to enable labeling with probes under mild conditions. It is supplied as a solid with high purity and is available in small milligram pack sizes or as a DMSO solution for biochemical applications.
  • High purity (98.0%) for reproducible results.
  • Hydrochloride salt, solid form for stable storage.
  • Soluble in DMSO (≈50 mg/mL); may require sonication.
  • Stable when stored sealed and kept away from moisture.
  • Suited for site-specific protein labeling in bacterial expression systems.