Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-amino-9-[(2S,3S,4R,5S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1H-purin-6-one | 26578-09-6 | MFCD00064103 | 98.0% | 283.24 | C10H13N5O5 | 5 MG
SDP

Catalog No. 5000221802
Encompass_Preferred

L-Guanosine is the L-enantiomer of the purine nucleoside guanosine, provided as a research-grade small molecule for biochemical and antiviral studies and for preparing supramolecular hydrogels. The material is characterized with defined physicochemical properties and recommended storage and handling conditions.

  • High purity: 98.0%.
  • Molecular formula: C10H13N5O5; molecular weight: 283.24.
  • Soluble in DMSO: approximately 100 mg/mL; ultrasonic assistance may be required.
  • Storage recommendations: store at 4°C; in solution store at -80°C for long-term or -20°C short-term.
  • Common applications: antiviral research, nucleoside chemistry, and hydrogel formation studies.

Catalog No. 50-002-21802 Supplier Medchemexpress LLC Supplier No. HY1381305MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Guanosine is the L-enantiomer of the purine nucleoside guanosine, provided as a research-grade small molecule for biochemical and antiviral studies and for preparing supramolecular hydrogels. The material is characterized with defined physicochemical properties and recommended storage and handling conditions.

  • High purity: 98.0%.
  • Molecular formula: C10H13N5O5; molecular weight: 283.24.
  • Soluble in DMSO: approximately 100 mg/mL; ultrasonic assistance may be required.
  • Storage recommendations: store at 4°C; in solution store at -80°C for long-term or -20°C short-term.
  • Common applications: antiviral research, nucleoside chemistry, and hydrogel formation studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.