Learn More
Apexbio Technology LLC 2-Amino-ATP(Synonyms: 2-NH2-ATP, 2-Aminoadenosine triphosphate, 2-Amino-adenosine-5'-triphosphate, 2-Amino-ATP (free acid)), 10ul (100 mM).
SDP
Catalog No. 5000060243
Encompass_Preferred
2-Amino-ATP is a modified nucleotide structurally related to ATP (adenosine triphosphate) featuring an amino substitution at the adenine ring s 2-position It acts as an ATP analog capable of competitively binding at the active sites of ATP-dependent enzymes potentially modulating their enzymatic activity and influencing downstream signaling pathways In research settings 2-Amino-ATP serves as a molecular tool to examine substrate specificity mechanistic aspects of ATP-binding enzymes (e g kinases and ATPases) and nucleotide-dependent receptor interactions facilitating studies of nucleotide signaling and cellular metabolism
Catalog No. 50-000-60243 Supplier Apexbio Technology LLC Supplier No. B79550.01
2-Amino-ATP is a modified nucleotide structurally related to ATP (adenosine triphosphate) featuring an amino substitution at the adenine ring s 2-position It acts as an ATP analog capable of competitively binding at the active sites of ATP-dependent enzymes potentially modulating their enzymatic activity and influencing downstream signaling pathways In research settings 2-Amino-ATP serves as a molecular tool to examine substrate specificity mechanistic aspects of ATP-binding enzymes (e g kinases and ATPases) and nucleotide-dependent receptor interactions facilitating studies of nucleotide signaling and cellular metabolism