Learn More
Polysciences, Inc. 2-Aminoethyl Methacrylate Hydrochloride; 10g; L.F. C6H 11 NO2.HCL; M.W. 166.6; > 98%; M.P. 121 deg. to 124 deg.C; CAS: 2420-94-2
SDP
Catalog No. NC9517435
Encompass_Preferred
2-Aminoethyl Methacrylate Hydrochloride; 10g; L.F. C6H 11 NO2.HCL; M.W. 166.6; > 98%; M.P. 121 deg. to 124 deg.C; CAS: 2420-94-2
Catalog No. NC9517435 Supplier Polysciences, Inc. Supplier No. 2100210
2-Aminoethyl Methacrylate Hydrochloride; 10g; L.F. C6H 11 NO2.HCL; M.W. 166.6; > 98%; M.P. 121 deg. to 124 deg.C; CAS: 2420-94-2