Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-m-cresol, 96%

Catalog No. p-3771182
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC398410050 5 g Glass bottle
AC398410250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC398410050 Supplier Thermo Scientific Chemicals Supplier No. 398410050
Only null left
Add to Cart

CAS: 2835-97-4 | C7H9NO | 123.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2835-97-4
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.16
MDL Number MFCD00075082
InChI Key FEDLEBCVFZMHBP-UHFFFAOYSA-N
Synonym 2-amino-m-cresol, phenol, 2-amino-3-methyl, 2-amino-3-methyl-phenol, 2-hydroxy-6-methylaniline, amino-m-cresol, ccris 8319, 2-azanyl-3-methyl-phenol, 2-hydroxy-6-methyl-aniline, acmc-1cp72, dsstox_cid_29117
PubChem CID 3014138
IUPAC Name 2-amino-3-methylphenol
SMILES CC1=CC=CC(O)=C1N

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 149.0°C to 152.0°C
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Linear Formula H2NC6H3(CH3)OH
Quantity 5 g
Formula Weight 123.15
Percent Purity 96%
Chemical Name or Material 2-Amino-m-cresol
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.