Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Amino-N-(14-azido-2,5-dioxo-9,12-dioxa-3,6-diazatetradecyl)acetamide | 2222277-20-3 | 95.0% | 345.35 g/mol | C12H23N7O5 | 10 MG
SDP

Catalog No. 5000204324
Encompass_Preferred

Gly-Gly-Gly-PEG2-azide is an azide-functional tri-glycine peptide with a PEG2 spacer used as an ADC linker and click-chemistry reagent for bioconjugation and conjugation of payloads to antibodies and other biomolecules via azide-alkyne cycloaddition.

  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted alkyne-azide cycloaddition (SPAAC).
  • CAS number 2222277-20-3.
  • Chemical formula C12H23N7O5.
  • Molecular weight 345.35 g/mol.
  • Purity 95.0%.
  • Available as powder and as 10 mM solution in DMSO; pack sizes include 5 mg, 10 mg, 50 mg (quote only), and 100 mg (quote only).
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (up to 6 months) or -20°C (up to 1 month).

Catalog No. 50-002-04324 Supplier Medchemexpress LLC Supplier No. HY15727510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gly-Gly-Gly-PEG2-azide is an azide-functional tri-glycine peptide with a PEG2 spacer used as an ADC linker and click-chemistry reagent for bioconjugation and conjugation of payloads to antibodies and other biomolecules via azide-alkyne cycloaddition.

  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted alkyne-azide cycloaddition (SPAAC).
  • CAS number 2222277-20-3.
  • Chemical formula C12H23N7O5.
  • Molecular weight 345.35 g/mol.
  • Purity 95.0%.
  • Available as powder and as 10 mM solution in DMSO; pack sizes include 5 mg, 10 mg, 50 mg (quote only), and 100 mg (quote only).
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (up to 6 months) or -20°C (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.