Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-amino-N-[[4-(5-bromopyrimidin-2-yl)oxy-3-chlorophenyl]carbamoyl]benzamide | 134742-26-0 | 99.2% | 462.68 g/mol | C18H13BrClN5O3 | 5 MG
SDP

Catalog No. 5000221632
Encompass_Preferred

NSC639828 is a small-molecule inhibitor of DNA polymerase α (IC50 = 70 μM) with reported antitumor activity. It is supplied as a solid research reagent suitable for enzymatic assays and preclinical cancer studies, with documented solubility and storage recommendations to support both in vitro and in vivo protocols.

  • Inhibits DNA polymerase α with IC50 of 70 μM.
  • High purity (≈99.2%) suitable for research applications.
  • Solid form: light yellow to green yellow powder.
  • Soluble in DMSO (100 mg/mL); in vivo formulation: 10% DMSO/90% corn oil (≥ 2.5 mg/mL).
  • Storage guidance: solid at -20°C, protect from light; in solution follow recommended temperatures and durations.
  • Offered in small pack sizes appropriate for laboratory experiments.

Catalog No. 50-002-21632 Supplier Medchemexpress LLC Supplier No. HY1453305MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NSC639828 is a small-molecule inhibitor of DNA polymerase α (IC50 = 70 μM) with reported antitumor activity. It is supplied as a solid research reagent suitable for enzymatic assays and preclinical cancer studies, with documented solubility and storage recommendations to support both in vitro and in vivo protocols.

  • Inhibits DNA polymerase α with IC50 of 70 μM.
  • High purity (≈99.2%) suitable for research applications.
  • Solid form: light yellow to green yellow powder.
  • Soluble in DMSO (100 mg/mL); in vivo formulation: 10% DMSO/90% corn oil (≥ 2.5 mg/mL).
  • Storage guidance: solid at -20°C, protect from light; in solution follow recommended temperatures and durations.
  • Offered in small pack sizes appropriate for laboratory experiments.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.