Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-amino-N-[4-[5-phenanthren-2-yl-3-(trifluoromethyl)pyrazol-1-yl]phenyl]acetamide | 742112-33-0 | MFCD12912272 | 98.7% | 460.45 | C26H19F3N4O | 100 MG
SDP

Catalog No. 5000241061
Encompass_Preferred

OSU-03012 (AR-12) is a blood-brain permeable inhibitor of 3-phosphoinositide-dependent protein kinase-1 (PDK-1) with a reported IC50 of 5 μM. It is supplied for research use and has documented activity in in vitro and in vivo studies relevant to oncology research.

  • Molecular weight: 460.45 g/mol.
  • Chemical formula: C26H19F3N4O.
  • Purity: 98.7% (typical).
  • Appearance: solid, white to off-white.
  • Storage: powder at -20°C (long term) or 4°C (short term); solutions at -80°C to -20°C.
  • Solubility: soluble in DMSO at ≥100 mg/mL.
  • Available forms: solid quantities and 10 mM DMSO solution for reconstitution.
  • Intended use: research use only, not for human or clinical use.

Catalog No. 50-002-41061 Supplier Medchemexpress LLC Supplier No. HY10547100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

OSU-03012 (AR-12) is a blood-brain permeable inhibitor of 3-phosphoinositide-dependent protein kinase-1 (PDK-1) with a reported IC50 of 5 μM. It is supplied for research use and has documented activity in in vitro and in vivo studies relevant to oncology research.

  • Molecular weight: 460.45 g/mol.
  • Chemical formula: C26H19F3N4O.
  • Purity: 98.7% (typical).
  • Appearance: solid, white to off-white.
  • Storage: powder at -20°C (long term) or 4°C (short term); solutions at -80°C to -20°C.
  • Solubility: soluble in DMSO at ≥100 mg/mL.
  • Available forms: solid quantities and 10 mM DMSO solution for reconstitution.
  • Intended use: research use only, not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.