Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-amino-N-[4-[5-phenanthren-2-yl-3-(trifluoromethyl)pyrazol-1-yl]phenyl]acetamide | 742112-33-0 | MFCD12912272 | 98.7% | 460.45 g/mol | C26H19F3N4O | 50 MG
SDP

Catalog No. 5000240043
Encompass_Preferred

OSU-03012 (AR-12) is a blood-brain permeable small-molecule inhibitor of 3-phosphoinositide-dependent kinase-1 (PDK1) used in preclinical research to modulate PDK1-dependent signaling and reduce cancer cell viability. It is supplied as a high-purity solid with characterized physicochemical and storage properties suitable for in vitro and in vivo studies.

  • Blood-brain permeable PDK1 inhibitor with reported cellular IC50 ~5 μM.
  • High purity solid, white to off-white.
  • Molecular weight 460.45 g/mol; formula C26H19F3N4O.
  • Soluble in DMSO ≥100 mg/mL (in vitro).
  • Recommended storage: powder -20°C (long-term) or 4°C (short-term).
  • Supplied in small-scale quantities suitable for research use.

Catalog No. 50-002-40043 Supplier Medchemexpress LLC Supplier No. HY1054750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

OSU-03012 (AR-12) is a blood-brain permeable small-molecule inhibitor of 3-phosphoinositide-dependent kinase-1 (PDK1) used in preclinical research to modulate PDK1-dependent signaling and reduce cancer cell viability. It is supplied as a high-purity solid with characterized physicochemical and storage properties suitable for in vitro and in vivo studies.

  • Blood-brain permeable PDK1 inhibitor with reported cellular IC50 ~5 μM.
  • High purity solid, white to off-white.
  • Molecular weight 460.45 g/mol; formula C26H19F3N4O.
  • Soluble in DMSO ≥100 mg/mL (in vitro).
  • Recommended storage: powder -20°C (long-term) or 4°C (short-term).
  • Supplied in small-scale quantities suitable for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.