Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-p-cresol Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. A1276500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1276500G 500 g
A127625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1276500G Supplier TCI America Supplier No. A1276500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2977-71-1
Molecular Formula C7H10ClNO
Molecular Weight (g/mol) 159.613
MDL Number MFCD00191600
InChI Key MZMUKGFYWNOPAU-UHFFFAOYSA-N
Synonym 2-Amino-4-methylphenol Hydrochloride, 2-Hydroxy-5-methylaniline Hydrochloride
PubChem CID 3014155
IUPAC Name 2-amino-4-methylphenol;hydrochloride
SMILES CC1=CC(=C(C=C1)O)N.Cl

Specifications

Quantity 500 g
Formula Weight 159.61
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-p-cresol Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.