Learn More
Medchemexpress LLC HY-I0501 500mg Medchemexpress, 2'-Aminoacetophenone CAS:551-93-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYI0501500MG
Encompass_Preferred
Medchemexpress, HY-I0501 500mg 2'-Aminoacetophenone CAS:551-93-9 2'-Aminoacetophenone is an aromatic compound containing a ketone substituted by one alkyl group, and a phenyl group. 2'-Aminoacetophenone can be used as a breath biomarker for the detection of Ps. Aeruginosa infections in the cystic fibrosis lung. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2759