Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminoazotoluene 97.0+%, TCI America™
SDP

Catalog No. T0261500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0261500G 500 g
T026125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0261500G Supplier TCI America Supplier No. T0261500G

Chemical Identifiers

CAS 97-56-3
Molecular Formula C14H15N3
Molecular Weight (g/mol) 225.295
MDL Number MFCD00007733
InChI Key PFRYFZZSECNQOL-UHFFFAOYSA-N
Synonym o-aminoazotoluene, fast garnet gbc base, solvent yellow 3, c.i. solvent yellow 3, toluazotoluidine, o-aminoazotoluol, fast oil yellow, somalia yellow r, fast yellow at, fat yellow b
PubChem CID 7340
IUPAC Name 2-methyl-4-[(2-methylphenyl)diazenyl]aniline
SMILES CC1=CC=CC=C1N=NC2=CC(=C(C=C2)N)C

Specifications

Melting Point 102°C
Quantity 500 g
Formula Weight 225.30
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminoazotoluene
EINECSNumber (5)-2299&(5)-5002
RTECSNumber XU8800000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.