Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Aminobenzaldehyde

Catalog No. A9628100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A9628100MG 100 mg
A96281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A9628100MG Supplier Sigma Aldrich Supplier No. A9628100MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Reactant for:
  • Preparation of quinoline derivatives as antiviral agents
  • Preparation of electroluminescent materials for oleds
  • Friedlander-type synthesis
  • Preparation of 2-tosylaminophenyl cyclopropylmethanols for gold-catalyzed cyclopropyl carbinol rearrangement
  • Benzyl c-h bond amination of arylmethylamines catalyzed by hydroxy-tempo
  • Silver-catalyzed aniline mediated cascade hydroamination/cycloaddition reactions

Specifications

CAS 529-23-7
Molecular Formula C7H7NO
Linear Formula C7H7NO
MDL Number MFCD00007709
Quantity 100 mg
Synonym 2-Formylaniline; Anthranilaldehyde; o-Aminobenzaldehyde
Molecular Weight (g/mol) 121.14
Percent Purity ≥98%
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.