Learn More
Selleck Chemical LLC 2-Aminobenzenesulfonamide S4998-25mg
SDP
Catalog No. 503112137
Encompass_Preferred
2-Aminobenzenesulfonamide (Orthanilamide O-Aminobenzenesulfonamide O-Sulfanilamide Benzenesulfonamide) is an inhibitor of Carbonic Anhydrase IX (CAIX) / Carbonic Anhydrase 9 (CA9)
Catalog No. 50-311-2137 Supplier Selleck Chemical LLC Supplier No. S499825MG
2-Aminobenzenesulfonamide (Orthanilamide O-Aminobenzenesulfonamide O-Sulfanilamide Benzenesulfonamide) is an inhibitor of Carbonic Anhydrase IX (CAIX) / Carbonic Anhydrase 9 (CA9)