Learn More
Medchemexpress LLC 2-aminobenzenesulfonic acid | 88-21-1 | MFCD00007705 | 99.9% | 173.19 g/mol | C6H7NO3S | 10 G
SDP
Catalog No. 5000302928
Encompass_Preferred
2-Aminobenzenesulfonic acid (orthanilic acid) is an aromatic sulfonic acid used as a versatile building block in organic synthesis. It is commonly used to prepare Schiff bases, to synthesize dyes and pharmaceutical intermediates, and to form coordination complexes with metals.
  • High purity (99.9%).
  • Solid reagent, easy to weigh and handle.
  • Useful for Schiff base formation and derivatization.
  • Suitable as an intermediate for dye and pharmaceutical synthesis.
  • Forms stable metal complexes for coordination chemistry.
  • Molecular formula C6H7NO3S; molecular weight 173.19 g/mol.
Catalog No. 50-003-02928 Supplier Medchemexpress LLC Supplier No. HYW01530210G
Add to Cart
Add to Cart
2-Aminobenzenesulfonic acid (orthanilic acid) is an aromatic sulfonic acid used as a versatile building block in organic synthesis. It is commonly used to prepare Schiff bases, to synthesize dyes and pharmaceutical intermediates, and to form coordination complexes with metals.
  • High purity (99.9%).
  • Solid reagent, easy to weigh and handle.
  • Useful for Schiff base formation and derivatization.
  • Suitable as an intermediate for dye and pharmaceutical synthesis.
  • Forms stable metal complexes for coordination chemistry.
  • Molecular formula C6H7NO3S; molecular weight 173.19 g/mol.