Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-aminobenzothiazole | 136-95-8 | MFCD00005785 | 99.7% | 150.21 g/mol | C7H6N2S | 1 ML
SDP

Catalog No. 5000300476
Encompass_Preferred

2-Aminobenzothiazole is an organic heterocycle used as a ligand, analytical standard, and intermediate in chemical research and synthesis. It is available both as a bulk solid and as a ready-to-use 10 mM solution in DMSO for laboratory workflows.

  • High purity (99.69% HPLC).
  • Molecular formula C7H6N2S.
  • Molecular weight 150.21 g/mol.
  • Supplied as 10 mM solution in DMSO and as bulk solid.
  • Suitable as ligand, ionophore, and drug intermediate.
  • Storage: room temperature protected from light; solutions stable at -80°C (2 years) or -20°C (1 year).

Catalog No. 50-003-00476 Supplier Medchemexpress LLC Supplier No. HYW01742410MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Aminobenzothiazole is an organic heterocycle used as a ligand, analytical standard, and intermediate in chemical research and synthesis. It is available both as a bulk solid and as a ready-to-use 10 mM solution in DMSO for laboratory workflows.

  • High purity (99.69% HPLC).
  • Molecular formula C7H6N2S.
  • Molecular weight 150.21 g/mol.
  • Supplied as 10 mM solution in DMSO and as bulk solid.
  • Suitable as ligand, ionophore, and drug intermediate.
  • Storage: room temperature protected from light; solutions stable at -80°C (2 years) or -20°C (1 year).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.