Learn More
Medchemexpress LLC 2-aminobenzothiazole | 136-95-8 | MFCD00005785 | 99.7% | 150.21 g/mol | C7H6N2S | 50 G

Supplier: Medchemexpress LLC HYW01742450G
2-Aminobenzothiazole is an aromatic heterocyclic research chemical used as a ligand in coordination chemistry, as a neutral ionophore for sensor membranes, and as a synthetic intermediate. Supplied for laboratory and analytical use, the product is accompanied by a certificate of analysis and safety data sheet confirming identity and assay.
- Purity confirmed by HPLC (99.69%).
- Molecular formula C7H6N2S and molecular weight 150.21 g/mol.
- Useful as a ligand, neutral ionophore, and synthetic intermediate.
- Supplied for research and analytical applications.
- COA and SDS available for traceability and compliance.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.