Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminobenzothiazole-6-carboxylic acid, 95%

Catalog No. AAH3151203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3151203 1 g
AAH3151206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3151203 Supplier Thermo Scientific Chemicals Supplier No. H3151203
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 93-85-6 | C8H6N2O2S | 194.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-85-6
Molecular Formula C8H6N2O2S
Molecular Weight (g/mol) 194.21
MDL Number MFCD00054180
InChI Key ZEAKWWWXCZMODH-UHFFFAOYSA-N
Synonym 2-aminobenzothiazole-6-carboxylic acid, 2-aminobenzo d thiazole-6-carboxylic acid, 6-benzothiazolecarboxylic acid, 2-amino, 2-amino-benzothiazole-6-carboxylic acid, 2-amino-6-benzothiazolecarboxylic acid, asischem b67429, zlchem 188, akos bb-7723, salor-int l481106-1ea, ksc487a1l
PubChem CID 66740
IUPAC Name 2-amino-1,3-benzothiazole-6-carboxylic acid
SMILES NC1=NC2=CC=C(C=C2S1)C(O)=O

Specifications

Melting Point >300°C
Quantity 1 g
Formula Weight 194.21
Percent Purity 95%
Chemical Name or Material 2-Aminobenzothiazole-6-carboxylic acid
Hazard Category H302
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 202-283-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.