Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminobenzothiazole, 97%

Catalog No. AC148811000
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC148811000 100 g Plastic bottle
AC148810050 5 g Glass bottle
AC148815000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC148811000 Supplier Thermo Scientific Chemicals Supplier No. 148811000
Only null left
Add to Cart

CAS: 136-95-8 | C7H6N2S | 150.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136-95-8
Molecular Formula C7H6N2S
Molecular Weight (g/mol) 150.2
MDL Number MFCD00005785
InChI Key UHGULLIUJBCTEF-UHFFFAOYSA-N
Synonym 2-aminobenzothiazole, 2-benzothiazolamine, benzo d thiazol-2-amine, benzothiazol-2-ylamine, 2-aminobenzthiazole, benzothiazole-2-ylamine, 2-iminobenzothiazoline, o-aminobenzothiazole, benzothiazole, 2-amino, 2-benzothiazolylamine
PubChem CID 8706
IUPAC Name 1,3-benzothiazol-2-amine
SMILES C1=CC=C2C(=C1)N=C(S2)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 129°C to 134°C
Color Beige to Gray
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Plastic bottle
Quantity 100 g
Beilstein 27, 182
Merck Index 15, 419
Solubility Information Solubility in water: sparingly soluble in water. Other solubilities: soluble in concentrated acids,freely soluble in alcohol,ether and chloroform
Formula Weight 150.2
Percent Purity 97%
Physical Form Powder or Flakes
Chemical Name or Material 2-Aminobenzothiazole, 97%
Show More Show Less
EINECSNumber 205-268-4
RTECSNumber DL1050000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.