Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminobenzothiazole Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. A121825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A121825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A121825G Supplier TCI America Supplier No. A121825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 94787-08-3
Molecular Formula C7H7ClN2S
Molecular Weight (g/mol) 186.657
MDL Number MFCD00142859
InChI Key UEQOZTASNGWDPC-UHFFFAOYSA-N
PubChem CID 3027575
IUPAC Name 1,3-benzothiazol-2-amine;hydrochloride
SMILES C1=CC=C2C(=C1)N=C(S2)N.Cl

Specifications

Melting Point 240°C
Color White
Quantity 25 g
Formula Weight 186.66
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminobenzothiazole Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.