Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminobenzotriazole 98.0+%, TCI America™
SDP

Catalog No. A1585100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1585100MG 100 mg
A15851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1585100MG Supplier TCI America Supplier No. A1585100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1614-11-5
Molecular Formula C6H6N4
Molecular Weight (g/mol) 134.142
MDL Number MFCD00724002
InChI Key YYZVLEJDGSWXII-UHFFFAOYSA-N
Synonym 2-Benzotriazolamine
PubChem CID 295514
IUPAC Name benzotriazol-2-amine
SMILES C1=CC2=NN(N=C2C=C1)N

Specifications

Melting Point 122°C
Color White-Yellow
Quantity 100 mg
Formula Weight 134.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminobenzotriazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.