Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminobenzotrifluoride 98.0+%, TCI America™
SDP

Catalog No. A0433500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0433500ML 500 mL
A043325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0433500ML Supplier TCI America Supplier No. A0433500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 88-17-5
Molecular Formula C7H6F3N
Molecular Weight (g/mol) 161.127
MDL Number MFCD00007718
InChI Key VBLXCTYLWZJBKA-UHFFFAOYSA-N
Synonym 2-trifluoromethyl aniline, 2-aminobenzotrifluoride, o-aminobenzotrifluoride, o-trifluoromethyl aniline, o-trifluoromethylaniline, benzenamine, 2-trifluoromethyl, 2-trifluoromethyl benzenamine, alpha,alpha,alpha-trifluoro-o-toluidine, 2-aminotrifluorotoluene, 2-aminotrifluoromethylbenzene
PubChem CID 6922
IUPAC Name 2-(trifluoromethyl)aniline
SMILES C1=CC=C(C(=C1)C(F)(F)F)N

Specifications

Boiling Point 171°C
Quantity 500 mL
UN Number 2942
Formula Weight 161.13
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Aminobenzotrifluoride
EINECSNumber (3)-0223
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.