Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Aminobenzyl alcohol

Catalog No. 12283125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-283-125G 25 g
12-283-110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-283-125G Supplier Sigma Aldrich Supplier No. 12283125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Aminobenzyl alcohol

Specifications

CAS 5344-90-1
Melting Point 81°C to 83°C (lit.)
Boiling Point 162°C (15 mmHg)
Molecular Formula C7H9NO
Linear Formula H2NC6H4CH2OH
MDL Number MFCD00007749
Quantity 25 g
Synonym 2-(Hydroxymethyl)aniline
Molecular Weight (g/mol) 123.15
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.