Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-aminobicyclohexyl

Catalog No. CDS013757 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS013757 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS013757 Supplier Sigma Aldrich Supplier No. CDS013757500MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1,1,1-trifluoro-5,5-dimethyl-2,4-hexanedione (tfdmhd) is a 1,3-diketone
  • It forms er(3+) complexes
  • These complexes shows thermally activated slow relaxation of magnetization under a zero direct-current (dc) field.1,1,1-trifluoro-5,5-dimethyl-2,4-hexanedione (tfdmhd) may be used in the synthesis of series of nno ketoimines bearing trifluoromethyl substituents, via schiff base condensation
  • It may be used in the synthesis of octahedral werner-type cobalt(ii) complexes
  • It was used in the preparation of d2-tfdmhd, by mixing with d2o

Specifications

CAS 6283-14-3
Molecular Formula C12H23N
Linear Formula C12H23N
MDL Number MFCD00021279
Quantity 5 mL
Molecular Weight (g/mol) 181.32
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.