Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-aminocarbonylphenylboronic acid

Catalog No. CDS006024 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS006024 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS006024 Supplier Sigma Aldrich Supplier No. CDS006024100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Ethyl trimethylacetate undergoes condensation with acetophenone catalyzed by phosphazene base to yield α-trimethylsilyloxy ester
  • Nci(f) and nci(nh2) mass spectra of a series of ethyl trimethylacetates have been studied

Specifications

CAS 380430-54-6
Molecular Formula C7H8BNO3
Linear Formula C7H8BNO3
MDL Number MFCD02179454
Quantity 100 g
Molecular Weight (g/mol) 164.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.