Learn More
Medchemexpress LLC 2-aminoethanesulfonic acid-13C2 | 70155-54-3 | MFCD01075566 | 99.0% | 127.13 g/mol | 13C2H7NO3S | 10 MG
SDP
Catalog No. 5000298966
Encompass_Preferred
Taurine-13C2 is a 13C-labeled form of taurine (2-aminoethanesulfonic acid) provided as a stable isotope standard for analytical and research use. It is supplied as a neat powder intended for use as an internal standard in mass spectrometry, isotope-dilution assays, and metabolic tracer studies.
  • 13C2-labeled taurine for use as an internal standard and tracer.
  • High isotopic enrichment and 99.0% purity.
  • Neat powder form, suitable for mass spectrometry and analytical workflows.
  • Small-mass package formats for isotope-dilution and tracer studies.
  • Molecular weight 127.13 g/mol; CAS number 70155-54-3.
Catalog No. 50-002-98966 Supplier Medchemexpress LLC Supplier No. HYB0351S110MG
Taurine-13C2 is a 13C-labeled form of taurine (2-aminoethanesulfonic acid) provided as a stable isotope standard for analytical and research use. It is supplied as a neat powder intended for use as an internal standard in mass spectrometry, isotope-dilution assays, and metabolic tracer studies.
  • 13C2-labeled taurine for use as an internal standard and tracer.
  • High isotopic enrichment and 99.0% purity.
  • Neat powder form, suitable for mass spectrometry and analytical workflows.
  • Small-mass package formats for isotope-dilution and tracer studies.
  • Molecular weight 127.13 g/mol; CAS number 70155-54-3.