Learn More
Medchemexpress LLC 2-aminoethanesulfonic acid-d4 | 342611-14-7 | 99.96% | 129.17 | C2H3D4NO3S | 10 MG
SDP
Catalog No. 5000297089
Encompass_Preferred
Taurine-d4 (2-aminoethanesulfonic acid-d4) is the deuterium-labeled analogue of taurine used as a stable-isotope standard in research and analytical workflows, including quantitative mass spectrometry and metabolic studies. It is supplied as a solid with a reported purity of 99.96% and a molecular weight of 129.17.
  • Deuterium-labeled internal standard for quantitative mass spectrometry.
  • High isotopic and chemical purity (99.96%).
  • Supplied as a solid powder suitable for analytical preparation.
  • Available in small measured quantities for trace-level analyses.
  • Appropriate for metabolic and biochemical research applications.
Catalog No. 50-002-97089 Supplier Medchemexpress LLC Supplier No. HYB0351S10MG
Taurine-d4 (2-aminoethanesulfonic acid-d4) is the deuterium-labeled analogue of taurine used as a stable-isotope standard in research and analytical workflows, including quantitative mass spectrometry and metabolic studies. It is supplied as a solid with a reported purity of 99.96% and a molecular weight of 129.17.
  • Deuterium-labeled internal standard for quantitative mass spectrometry.
  • High isotopic and chemical purity (99.96%).
  • Supplied as a solid powder suitable for analytical preparation.
  • Available in small measured quantities for trace-level analyses.
  • Appropriate for metabolic and biochemical research applications.